siRNA Details
Gene Symbol: Dcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007831.3 | NP_031857.2 | 9 | 2133 | ||
GenBank mRNA | AK157715.1 | - | 9 | 2082 | ||
X85788.1 | - | 6 | 1683 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007831.3 | NP_031857.2 | 9 | 2133 | ||
GenBank mRNA | AK157715.1 | - | 9 | 2082 | ||
X85788.1 | - | 6 | 1683 |
Set Membership: |
![]() |