siRNA Details
Gene Symbol: Dcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007831.3 | NP_031857.2 | 8, 9 | 2050 | ||
GenBank mRNA | AK157715.1 | - | 8, 9 | 1999 | ||
X85788.1 | - | 5, 6 | 1600 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
