siRNA Details
Gene Symbol: Pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025273.4 | NP_079549.1 | 2 | 180 | ||
GenBank mRNA | AK007401.1 | - | 2 | 152 | ||
BC024354.1 | - | 2 | 113 | 5052788 | ||
BY294059.1 | - | 2 | 168 | |||
M83741.1 | - | 1 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
