siRNA Details
Gene Symbol: Ddit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290183.1 | NP_001277112.1 | 2, 3 | 266 | ||
NM_007837.4 | NP_031863.3 | 3, 4 | 314 | |||
GenBank mRNA | AA929408.1 | - | 2, 3 | 245 | ||
AK028120.1 | - | 3, 4 | 311 | |||
BC013718.1 | - | 3, 4 | 309 | 4012433 | ||
BY121546.1 | - | 3, 4 | 314 | |||
X67083.1 | - | 2, 3 | 221 |

More Information
Other Species Targeted

Gene Ontology Categories:
