siRNA Details
Gene Symbol: Ddost | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007838.2 | NP_031864.2 | 4 | 565 | ||
GenBank mRNA | AK077651.1 | - | 4 | 428 | ||
AK081542.1 | - | 4 | 457 | |||
AK084267.1 | - | 4 | 449 | |||
AK148095.1 | - | 4 | 446 | |||
AK150683.1 | - | 4 | 456 | |||
AK182353.1 | - | 2 | 101 | |||
AK196638.1 | - | 2 | 134 | |||
AK200087.1 | - | 2 | 126 | |||
AK201961.1 | - | 2 | 101 | |||
AK204319.1 | - | 2 | 101 | |||
BC068132.1 | - | 4 | 565 | 6808198 | ||
D89063.1 | - | 4 | 567 |

More Information
Other Species Targeted

Gene Ontology Categories:
