siRNA Details
Gene Symbol: Dhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007842.2 | NP_031868.2 | 17 | 1991 | ||
GenBank mRNA | AK135603.1 | - | 12 | 1306 | ||
AK141824.1 | - | 17 | 1986 | |||
AY512925.1 | - | 13 | 1447 | |||
BC089159.1 | - | 17 | 1980 | |||
U91922.1 | - | 17 | 1948 | |||
U92080.1 | - | 6 | 686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
