siRNA Details
Gene Symbol: Defb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007843.3 | NP_031869.1 | 2 | 180 | ||
GenBank mRNA | AI956736.1 | - | 2 | 246 | ||
AK142544.1 | - | 2 | 161 | |||
BC024380.1 | - | 2 | 136 | 4975536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
