siRNA Details
Gene Symbol: Defa-rs10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007845.3 | NP_031871.1 | 175 | |||
GenBank mRNA | BC119615.1 | - | 175 | 40061719 | ||
BC119616.1 | - | 175 | 40061721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
