siRNA Details
Gene Symbol: Twist2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007855.2 | NP_031881.1 | 2 | 952 | ||
GenBank mRNA | AK008388.1 | - | 2 | 690 | ||
AK008411.1 | - | 2 | 689 | |||
AK011180.1 | - | 2 | 968 | |||
AK011861.1 | - | 2 | 969 | |||
BC090636.1 | - | 2 | 957 | 30942245 | ||
U36384.1 | - | 2 | 952 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
