siRNA Details
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007861.5 | NP_031887.2 | 7, 8 | 715 | ||
GenBank mRNA | AK117104.1 | - | 7, 8 | 659 | ||
AK135807.1 | - | 7, 8 | 658 | |||
AK136193.1 | - | 7, 8 | 659 | |||
AK153399.1 | - | 7, 8 | 659 | |||
AK167885.1 | - | 7, 8 | 659 | |||
AK168157.1 | - | 7, 8 | 680 | |||
AK168875.1 | - | 7, 8 | 600 | |||
AK193031.1 | - | 1, 2 | 115 | |||
BC003368.1 | - | 7, 8 | 642 | 3589344 | ||
U73445.1 | - | 7, 8 | 692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
