siRNA Details
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007864.3 | NP_031890.1 | 3, 4 | 474 | ||
GenBank mRNA | BI738861.1 | - | 3, 4 | 218 | ||
D50621.1 | - | 3, 4 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
