siRNA Details
Gene Symbol: HDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004494.2 | NP_004485.1 | 1, 2 | 416 | ||
GenBank mRNA | AK298342.1 | - | 1, 2 | 166 | ||
AK315514.1 | - | 1, 2 | 185 | |||
BC018991.1 | - | 1, 2 | 267 | 3503321 | ||
D16431.1 | - | 1, 2 | 409 | |||
DR159090.1 | - | 1, 2 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
