siRNA Details
Gene Symbol: Dmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278226.1 | NP_001265155.1 | 5 | 451 | ||
NM_010059.3 | NP_034189.1 | 5 | 451 | |||
GenBank mRNA | AK076777.1 | - | 5 | 442 | ||
AK169570.1 | - | 5 | 451 | |||
BC116767.1 | - | 5 | 336 | 40126086 | ||
BC119081.1 | - | 5 | 336 | 8733830 | ||
D58419.1 | - | 5 | 436 | |||
D64107.1 | - | 5 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
