siRNA Details
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190817.1 | NP_001177746.1 | 5 | 942 | ||
NM_007869.3 | NP_031895.1 | 5 | 942 | |||
GenBank mRNA | AK133582.1 | - | 5 | 924 | ||
AK144228.1 | - | 5 | 898 | |||
AK164032.1 | - | 2 | 1564 | |||
BC080300.1 | - | 5 | 923 | |||
L16953.1 | - | 5 | 619 |
