siRNA Details
Gene Symbol: Dnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301737.1 | NP_001288666.1 | 12 | 1555 | ||
NM_010065.3 | NP_034195.2 | 12 | 1555 | |||
GenBank mRNA | AK011651.1 | - | 12 | 1522 | ||
AK144142.1 | - | 12 | 1506 | |||
AK220483.1 | - | 12 | 1497 | |||
BC034679.1 | - | 12 | 1509 | 4506865 | ||
BC058623.1 | - | 12 | 1489 | 5706803 | ||
BQ179370.1 | - | 6 | 480 | |||
L31395.1 | - | 12 | 1452 | |||
L31396.1 | - | 7 | 724 | |||
L31397.1 | - | 12 | 1508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
