siRNA Details
Gene Symbol: Dnmt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003960.4 | NP_001003960.2 | 7, 8 | 1087 | ||
NM_001003961.4 | NP_001003961.2 | 7, 8 | 1087 | |||
NM_001003963.4 | NP_001003963.2 | 7, 8 | 1087 | |||
NM_001122997.2 | NP_001116469.1 | 6, 7 | 972 | |||
NM_001271744.1 | NP_001258673.1 | 6, 7 | 972 | |||
NM_001271745.1 | NP_001258674.1 | 6, 7 | 972 | |||
NM_001271746.1 | NP_001258675.1 | 6, 7 | 972 | |||
NM_001271747.1 | NP_001258676.1 | 6, 7 | 972 | |||
NM_010068.5 | NP_034198.3 | 7, 8 | 1087 | |||
GenBank mRNA | AF068626.2 | - | 7, 8 | 942 | ||
AF068627.2 | - | 7, 8 | 942 | |||
AF068628.2 | - | 7, 8 | 942 | |||
AF151969.1 | - | 7, 8 | 1085 | |||
AF151970.1 | - | 7, 8 | 1085 | |||
AF151971.1 | - | 7, 8 | 1085 | |||
AF151972.1 | - | 7, 8 | 1085 | |||
AF151973.1 | - | 6, 7 | 970 | |||
AF151974.1 | - | 6, 7 | 970 | |||
AF151975.1 | - | 6, 7 | 970 | |||
AF151976.1 | - | 6, 7 | 970 | |||
AY078427.1 | - | 7, 8 | 942 | |||
BC105677.1 | - | 6, 7 | 804 | 40048574 | ||
BC105922.1 | - | 6, 7 | 804 | 40048584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
