siRNA Details
Gene Symbol: Doc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007873.2 | NP_031899.2 | 7, 8 | 1175 | ||
GenBank mRNA | BC067030.1 | - | 7, 8 | 1175 | 30534150 | |
D85037.1 | - | 7, 8 | 1167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
