siRNA Details
Gene Symbol: Dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010073.2 | NP_034203.1 | 4 | 371 | ||
GenBank mRNA | AK002468.1 | - | 4 | 355 | ||
AK004241.1 | - | 4 | 346 | |||
BC008256.1 | - | 4 | 371 | 2648752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
