siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159543.1 | NP_001153015.1 | 16 | 1762 | ||
NM_010074.3 | NP_034204.1 | 16 | 1762 | |||
GenBank mRNA | AK085370.1 | - | 16 | 1345 | ||
AK153939.1 | - | 16 | 1376 | |||
AK163085.1 | - | 16 | 1767 | |||
AK164775.1 | - | 16 | 1345 | |||
BC022183.1 | - | 17 | 1624 | 3982742 | ||
X58384.1 | - | 16 | 1439 |

More Information
Other Species Targeted

Gene Ontology Categories:
