siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159543.1 | NP_001153015.1 | 15 | 1678 | ||
NM_010074.3 | NP_034204.1 | 15 | 1678 | |||
GenBank mRNA | AK085370.1 | - | 15 | 1261 | ||
AK153939.1 | - | 15 | 1292 | |||
AK163085.1 | - | 15 | 1683 | |||
AK164775.1 | - | 15 | 1261 | |||
BC022183.1 | - | 16 | 1540 | 3982742 | ||
X58384.1 | - | 15 | 1355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
