siRNA Details
Gene Symbol: Drd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032054.1 | - | 1 | 1120 | ||
AK045456.1 | - | 1 | 1309 | |||
BC138059.1 | - | 1 | 1177 | 8861078 | ||
BC138060.1 | - | 1 | 1177 | 8861079 | ||
S77992.1 | - | 1 | 17 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
