siRNA Details
Gene Symbol: Dsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013505.3 | NP_038533.1 | 12 | 2097 | ||
GenBank mRNA | AK028604.1 | - | 13 | 2137 | ||
AK028636.1 | - | 12 | 2098 | |||
AK030333.1 | - | 12 | 2098 | |||
AK048547.1 | - | 12 | 2098 | |||
AK172168.1 | - | 12 | 1932 | |||
BC004663.1 | - | 6 | 984 | |||
BC057867.1 | - | 12 | 1877 | 5327748 | ||
L33779.1 | - | 12 | 1896 | |||
X73885.1 | - | 5 | 815 |
