siRNA Details
Gene Symbol: Slc26a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007885.2 | NP_031911.1 | 4 | 2262 | ||
GenBank mRNA | AK052942.1 | - | 4 | 2263 | ||
AK163974.1 | - | 4 | 2153 | |||
AK165805.1 | - | 2 | 1569 | |||
BC028345.1 | - | 4 | 2139 | 4014785 | ||
D42049.1 | - | 4 | 2147 |
