siRNA Details
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291105.1 | NP_001278034.1 | 3 | 482 | ||
NM_007891.5 | NP_031917.1 | 3 | 526 | |||
GenBank mRNA | AF483514.1 | - | 3 | 368 | ||
AF483515.1 | - | 3 | 368 | |||
AK017841.1 | - | 3 | 474 | |||
AK153968.1 | - | 3 | 491 | |||
AK220226.1 | - | 3 | 223 | |||
BC052160.1 | - | 3 | 468 | 6330425 | ||
L21973.1 | - | 3 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
