siRNA Details
Gene Symbol: E2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289920.1 | NP_001276849.1 | 4 | 709 | ||
NM_010093.3 | NP_034223.1 | 4 | 1227 | |||
GenBank mRNA | AF015948.1 | - | 4 | 529 | ||
AK129048.1 | - | 3 | 425 | |||
AK133828.1 | - | 4 | 1229 | |||
AK165892.1 | - | 4 | 710 | |||
BC024125.1 | - | 4 | 466 | |||
BC059262.1 | - | 4 | 626 |

More Information
Other Species Targeted

Gene Ontology Categories:
