siRNA Details
Gene Symbol: Ear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007894.2 | NP_031920.1 | 399 | |||
GenBank mRNA | AY316149.1 | - | 399 | |||
BC117060.1 | - | 2 | 353 | 40126379 | ||
U72032.1 | - | 1 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
