siRNA Details
Gene Symbol: Ear2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007895.2 | NP_031921.1 | 2 | 645 | ||
GenBank mRNA | AF306665.1 | - | 2 | 645 | ||
BC065391.1 | - | 2 | 616 | 3464790 | ||
BC094626.1 | - | 2 | 612 | 1515361 |
Gene Symbol: Ear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007894.2 | NP_031920.1 | 639 | |||
GenBank mRNA | AY316149.1 | - | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
