siRNA Details
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290709.1 | NP_001277638.1 | 6 | 620 | ||
NM_001290710.1 | NP_001277639.1 | 6 | 620 | |||
NM_001290711.1 | NP_001277640.1 | 6 | 739 | |||
NM_007897.3 | NP_031923.1 | 6 | 620 | |||
GenBank mRNA | AK031239.1 | - | 6 | 615 | ||
AK035781.1 | - | 6 | 503 | |||
AK051438.1 | - | 6 | 619 | |||
AK137389.1 | - | 6 | 661 | |||
AK139021.1 | - | 6 | 618 | |||
AK141175.1 | - | 6 | 620 | |||
BC139362.1 | - | 6 | 581 | 8862384 | ||
BC139364.1 | - | 6 | 581 | 8862386 | ||
L12147.1 | - | 6 | 586 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053820, XM_008767631, XM_006246118, XM_006246117, XM_008767634, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
