siRNA Details
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037987.3 | NP_001033076.1 | 5 | 969 | ||
NM_010103.4 | NP_034233.1 | 4 | 939 | |||
GenBank mRNA | AF031524.1 | - | 5 | 1077 | ||
AF031525.1 | - | 5 | 1008 | |||
AK036117.1 | - | 5 | 930 | |||
AK046885.1 | - | 5 | 970 | |||
AK081003.1 | - | 4 | 771 | |||
AK084215.1 | - | 5 | 824 | |||
AK135220.1 | - | 5 | 933 | |||
AK139093.1 | - | 5 | 931 | |||
AK141907.1 | - | 5 | 953 | |||
AK157810.1 | - | 5 | 954 | |||
BC056386.1 | - | 5 | 869 | 6854497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
