siRNA Details
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037987.3 | NP_001033076.1 | 5 | 919 | ||
NM_010103.4 | NP_034233.1 | 4 | 889 | |||
GenBank mRNA | AF031524.1 | - | 5 | 1027 | ||
AF031525.1 | - | 5 | 958 | |||
AK036117.1 | - | 5 | 880 | |||
AK046885.1 | - | 5 | 920 | |||
AK081003.1 | - | 4 | 721 | |||
AK084215.1 | - | 5 | 774 | |||
AK135220.1 | - | 5 | 883 | |||
AK139093.1 | - | 5 | 881 | |||
AK141907.1 | - | 5 | 903 | |||
AK157810.1 | - | 5 | 904 | |||
BC056386.1 | - | 5 | 819 | 6854497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
