siRNA Details
Gene Symbol: Edn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010104.3 | NP_034234.1 | 4 | 1049 | ||
GenBank mRNA | AB081657.1 | - | 4 | 435 | ||
AK040778.1 | - | 4 | 707 | |||
AK152647.1 | - | 4 | 700 | |||
AK153437.1 | - | 4 | 704 | |||
BC029547.1 | - | 4 | 843 | 5059522 | ||
D43775.1 | - | 4 | 760 | |||
U35233.1 | - | 4 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
