siRNA Details
Gene Symbol: Ednrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136061.2 | NP_001129533.1 | 4 | 1037 | ||
NM_001276296.1 | NP_001263225.1 | 3 | 851 | |||
NM_007904.4 | NP_031930.1 | 3 | 851 | |||
GenBank mRNA | AK076426.1 | - | 3 | 845 | ||
AK082103.1 | - | 4 | 838 | |||
AK083415.1 | - | 4 | 734 | |||
AK085165.1 | - | 3 | 842 | |||
AK085532.1 | - | 3 | 847 | |||
AK192059.1 | - | 3 | 304 | |||
AK195461.1 | - | 3 | 300 | |||
AK198739.1 | - | 3 | 284 | |||
AK199447.1 | - | 3 | 276 | |||
AK202909.1 | - | 3 | 248 | |||
AK204434.1 | - | 3 | 270 | |||
AK211790.1 | - | 3 | 311 | |||
AK215891.1 | - | 3 | 177 | |||
AK219164.1 | - | 3 | 295 | |||
BC026553.1 | - | 4 | 963 | 4971909 | ||
U32329.1 | - | 3 | 829 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
