siRNA Details
Gene Symbol: Eed | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021876.3 | NP_068676.1 | 3, 4 | 816 | ||
GenBank mRNA | AK077664.1 | - | 3, 4 | 401 | ||
AK087924.1 | - | 3, 4 | 378 | |||
AK131976.1 | - | 3, 4 | 383 | |||
AK133890.1 | - | 3, 4 | 377 | |||
BC012966.1 | - | 3, 4 | 815 | 3991086 | ||
U78103.1 | - | 3, 4 | 818 | |||
U97675.1 | - | 3, 4 | 823 |
