siRNA Details
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010112.4 | NP_034242.2 | 4 | 2433 | ||
GenBank mRNA | AK030321.1 | - | 4 | 1560 | ||
AK163191.1 | - | 4 | 1474 | |||
BC005438.1 | - | 4 | 1502 | 3487600 | ||
BC031543.1 | - | 4 | 1228 | 3669513 | ||
U28728.1 | - | 4 | 1457 | |||
U57686.1 | - | 4 | 1157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
