siRNA Details
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199494.1 | NP_001186423.1 | 9 | 944 | ||
NM_007915.5 | NP_031941.1 | 9 | 828 | |||
GenBank mRNA | AK145018.1 | - | 9 | 905 | ||
AK147776.1 | - | 9 | 857 | |||
AK159927.1 | - | 9 | 886 | |||
AK172233.1 | - | 9 | 908 | |||
U41751.2 | - | 9 | 832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
