siRNA Details
Gene Symbol: Eif2ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010121.2 | NP_034251.2 | 10 | 1935 | ||
GenBank mRNA | AF076681.1 | - | 10 | 1927 | ||
AK139679.1 | - | 8 | 1257 | |||
BC054809.1 | - | 10 | 1947 | 6839221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
