siRNA Details
Gene Symbol: Ddx19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007916.2 | NP_031942.2 | 5, 6 | 497 | ||
GenBank mRNA | AK049467.1 | - | 5, 6 | 468 | ||
AK146548.1 | - | 5, 6 | 458 | |||
BC011270.1 | - | 5, 6 | 469 | 3674608 | ||
L25125.1 | - | 5, 6 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
