siRNA Details
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123037.2 | NP_001116509.1 | 9 | 1199 | ||
NM_001123038.2 | NP_001116510.1 | 10 | 1339 | |||
NM_013506.3 | NP_038534.2 | 10 | 1339 | |||
GenBank mRNA | AK076509.1 | - | 10 | 1050 | ||
AK088669.1 | - | 10 | 1051 | |||
AK132969.1 | - | 10 | 1050 | |||
AK165898.1 | - | 9 | 910 | |||
BC094422.1 | - | 10 | 1035 | 6808404 | ||
CK623185.1 | - | 5 | 446 | |||
X12507.1 | - | 10 | 1051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
