siRNA Details
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123038.2 | NP_001116510.1 | 4, 5 | 675 | ||
NM_013506.3 | NP_038534.2 | 4, 5 | 675 | |||
GenBank mRNA | AK076509.1 | - | 4, 5 | 386 | ||
AK088669.1 | - | 4, 5 | 387 | |||
AK132969.1 | - | 4, 5 | 386 | |||
BC094422.1 | - | 4, 5 | 371 | 6808404 | ||
CF723811.1 | - | 4, 5 | 567 | |||
X12507.1 | - | 4, 5 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
