siRNA Details
Gene Symbol: Eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010124.2 | NP_034254.1 | 2 | 407 | ||
GenBank mRNA | AK133912.1 | - | 2 | 295 | ||
AK148374.1 | - | 446 | ||||
AK210066.1 | - | 1 | 145 | |||
AK216468.1 | - | 1 | 145 | |||
AK217241.1 | - | 1 | 145 | |||
BC015082.1 | - | 3 | 388 | 4218820 | ||
BC119539.2 | - | 3 | 371 | 40056467 | ||
U75530.1 | - | 2 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
