siRNA Details
Gene Symbol: Elk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007923.2 | NP_031949.2 | 2 | 543 | ||
GenBank mRNA | AK054075.1 | - | 2 | 835 | ||
AK156537.1 | - | 2 | 541 | |||
BC144844.1 | - | 2 | 293 | 9053392 | ||
BC145795.1 | - | 2 | 293 | 40131098 | ||
FN434116.1 | - | 1 | 140 | |||
FN434117.1 | - | 1 | 140 | |||
FN434118.1 | - | 1 | 140 | |||
Z36885.1 | - | 3 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
