siRNA Details
Gene Symbol: En1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010133.2 | NP_034263.2 | 1 | 863 | ||
GenBank mRNA | AK140408.1 | - | 1 | 726 | ||
AK140634.1 | - | 1 | 937 | |||
L12703.1 | - | 1 | 669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
