siRNA Details
Gene Symbol: Endog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007931.1 | NP_031957.1 | 2 | 601 | ||
GenBank mRNA | AB012108.1 | - | 2 | 601 | ||
AK144459.1 | - | 2 | 633 | |||
BC030177.1 | - | 2 | 610 | 5029633 | ||
X99394.1 | - | 2 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
