siRNA Details
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136062.2 | NP_001129534.1 | 4, 5 | 342 | ||
NM_001276285.1 | NP_001263214.1 | 4, 5 | 354 | |||
NM_007933.3 | NP_031959.1 | 4, 5 | 454 | |||
GenBank mRNA | AK002485.1 | - | 4, 5 | 314 | ||
BC013460.1 | - | 3, 4 | 326 | 4217678 | ||
BY115250.1 | - | 4, 5 | 327 | |||
CT010193.1 | - | 3, 4 | 248 | |||
M20745.1 | - | 1, 2 | 74 | |||
X57747.1 | - | 4, 5 | 305 | |||
X62667.1 | - | 3, 4 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
