siRNA Details
Gene Symbol: Eomes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164789.1 | NP_001158261.1 | 6 | 2075 | ||
NM_010136.3 | NP_034266.2 | 6 | 2132 | |||
GenBank mRNA | AB031037.1 | - | 6 | 2047 | ||
AK089817.1 | - | 6 | 1692 | |||
AK134951.1 | - | 1287 | ||||
AK143454.1 | - | 6 | 1453 | |||
BC094319.1 | - | 6 | 1977 | 5721456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
