siRNA Details
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010137.3 | NP_034267.3 | 13 | 2553 | ||
GenBank mRNA | AF045160.1 | - | 13 | 2317 | ||
AK136911.1 | - | 13 | 2557 | |||
AK141185.1 | - | 13 | 2288 | |||
AK158430.1 | - | 13 | 2717 | |||
BC057870.1 | - | 13 | 2275 | 5032291 | ||
D89787.1 | - | 13 | 2515 | |||
U81983.1 | - | 13 | 2236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
