siRNA Details
Gene Symbol: Epb4.1l4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013512.2 | NP_038540.2 | 14 | 1876 | ||
GenBank mRNA | AK140485.1 | - | 13 | 1560 | ||
AK141236.1 | - | 2 | 46 | |||
BC007166.1 | - | 14 | 1600 | 3490061 | ||
BC013557.1 | - | 14 | 1600 | 3490326 | ||
BC089158.1 | - | 13 | 1448 | |||
D28818.1 | - | 13 | 1144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
