siRNA Details
Gene Symbol: Epb4.1l4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013512.2 | NP_038540.2 | 3 | 915 | ||
GenBank mRNA | AK140485.1 | - | 2 | 599 | ||
AK161182.1 | - | 3 | 675 | |||
BC007166.1 | - | 3 | 639 | 3490061 | ||
BC011099.1 | - | 2 | 234 | |||
BC013557.1 | - | 3 | 639 | 3490326 | ||
BC089158.1 | - | 2 | 506 | |||
D28818.1 | - | 2 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
