siRNA Details
Gene Symbol: Epha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007938.2 | NP_031964.2 | 10 | 2391 | ||
GenBank mRNA | AK032436.1 | - | 6 | 636 | ||
AK077099.1 | - | 4 | 401 | |||
BC141090.1 | - | 10 | 1907 | 9055702 | ||
BC145362.1 | - | 11 | 2027 | |||
U58332.1 | - | 10 | 2107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
