siRNA Details
Gene Symbol: Epha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007938.2 | NP_031964.2 | 17, 18 | 3564 | ||
GenBank mRNA | AK032436.1 | - | 13, 14 | 1809 | ||
BC141090.1 | - | 17, 18 | 3080 | 9055702 | ||
BC145362.1 | - | 18, 19 | 3200 | |||
U58332.1 | - | 17, 18 | 3280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
